missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Wide mouth, Short form DIN thread
Square clear soda lime glass bottles, wide mouth.
21.70 € - 105.00 €
Specifications
For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
---|---|
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
13187374
|
DWK Life Sciences
238203607 |
250 mL |
53.60 €
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
13124354
|
DWK Life Sciences
238204406 |
500 mL |
65.90 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
13107444
|
DWK Life Sciences
238205402 |
1000 mL | N/A | ||||||
12891004
|
DWK Life Sciences
238202405 |
100 mL |
30.65 €
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10787945
|
DWK Life Sciences
238201709 |
50 mL | N/A | ||||||
Description
Square clear soda lime (Type 3) glass bottles, wide mouth and short form DIN thread.
- Square shape for efficient packing
- Wide bottle neck allows easy access
Specifications
Storage of samples and reagents; Sample collection; Transport and shipping | |
Soda Lime Glass | |
10/Pk. | |
Not Autoclavable |