Learn More
Lenz Laborglasintrumente Screwthread Tubes For Glassblowers
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
22.00 € - 190.00 €
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
10353661
|
Lenz Laborglasintrumente
01300018 |
18 GL |
22.00 €
Pack of 10 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
10048440
|
Lenz Laborglasintrumente
01300045 |
45 GL |
190.00 €
Pack of 10 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10008420
|
Lenz Laborglasintrumente
01300025 |
25 GL |
36.00 €
Pack of 10 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
Description
- Acc. to DIN 12216 and in addition to standard
- With DIN thread (GL)
- Material: DURAN™
Specifications
Borosilicate Glass (3.3) | |
10/Pk. |