missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SDHB/CEN1p FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Description
Includes
DAPI Counterstain (1500ng/mL) 125μL
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Label or Dye | FITC, Texas Red |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) |
| Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
| Location | 1p36.13, 1p13.3 |
| Probe | SDHB, CEN1p |
| Product Type | SDHB/CEN1p FISH Probe |
| Quantity | 100 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction