missing translation for 'onlineSavingsMsg'
Learn More
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™ Product Code.: 10607443

SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™

Product Code. 10607443
1 L, Nalgene Poly Bottle
Click to view available options
Quantity:
200 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
This item is not returnable. View return policy

Product Code. 10607443

Brand: Fisher BioReagents BP13111

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis

  • DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
  • Reduce preparation time by using this premade SDS solution
  • SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
  • SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3, 7732-18-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
Color Colorless
Physical Form Liquid
Boiling Point 100°C
Density 1.01g/cm³
Concentration 19.5 to 20.5%
Quantity 1 L
Identification Pass Test
Packaging Nalgene Poly Bottle
Assay Percent Range (as Fatty Alcohol Sulfate): ≥99%
Linear Formula CH3(CH2)10CH2OSO3Na
Merck Index 15, 8768
IUPAC Name sodium dodecyl sulfate
Formula Weight 288.38
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Sodium Dodecyl Sulfate, 20% Solution
Show More Show Less
compliance-icons
Product Identifier
  • Sodium dodecyl sulfate, 10% solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.