missing translation for 'onlineSavingsMsg'
Learn More

SDZ 220-581, Tocris Bioscience™

Product Code. 18035942
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Competitive NMDA antagonist

Competitive NMDA receptor antagonist (pKi = 7.7). Centrally active following oral administration (ED50 < 3.2 mg/kg for protection against MES-induced seizures).

Chemical Identifiers

CAS 174575-17-8
Molecular Formula C16H17ClNO5P
Molecular Weight (g/mol) 369.738
InChI Key VBRJFXSFCYEZMQ-HNNXBMFYSA-N
Synonym 2s-2-amino-3-3-2-chlorophenyl-5-phosphonomethyl phenyl propanoic acid, 2s-2-amino-3-2'-chloro-5-phosphonomethyl-1,1'-biphenyl-3-yl propanoic acid, tocris-1250, biomol-nt_000193, alpha-amino-2'-chloro-5-phosphonomethyl 1,1'-biphenyl-3-propanoic acid, 1,1'-biphenyl-3-propanoic acid, alpha-amino-2'-chloro-5-phosphonomethyl-, s, s-?-amino-2'-chloro-5-phosphonomethyl 1,1'-biphenyl-3-propanoic acid, s-alpha-amino-2'-chloro-5-phosphonomethyl-1,1'-biphenyl-3-propanoic acid, s-alpha-amino-2'-chloro-5-phosphonomethyl 1,1'-biphenyl-3-propanoic acid
PubChem CID 128019
IUPAC Name (2S)-2-amino-3-[3-(2-chlorophenyl)-5-(phosphonomethyl)phenyl]propanoic acid
SMILES C1=CC=C(C(=C1)C2=CC(=CC(=C2)CP(=O)(O)O)CC(C(=O)O)N)Cl

Specifications

Quantity 10 mg
Formula Weight 369.74
Percent Purity >98%
Chemical Name or Material SDZ 220-581
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'