Learn More
Hoefer™ SE640 Wide-Mini Vertical Electrophoresis Unit
Wide-mini system, ideal for rapid screening of multiple samples
Brand:  Hoefer™ SE640
 View more versions of this product
Product Code. 11985408
  • 1275.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Includes:
  • Basic package: Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, Spacer-Mate alignment template, Wonder Wedge plate separation tool, and Gel Seal.
  • Complete package: Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, Spacer-Mate alignment template, Wonder Wedge plate separation tool, Gel Seal, two 15-well 1.5mm combs and two sets of 1.5mm thick (2cm wide) spacers.
Description
Description
The Hoefer Wide-Mini Vertical Electrophoresis unit offers extra wide format ideal for quick, high throughput runs. With accessory divider plate you can complete a run of up to 112 samples in as little as 40 minutes.
  • Maximum glass plate size 18 x 8 cm, permits rapid screening of up to 112 samples at one time
  • Design permits gels to be surrounded uniformly by buffer so heat dissipation is efficient; bands are sharp and lanes are straight
  • Accommodates denaturing or native polyacrylamide gels
  • Run up to four second dimension gels suing IEF strips or tube gels
  • Ideal for optimizing 2DE protocols
  • Storage temperature: +15°C to 30°C
Specifications
Specifications
EN61010-1, CSA C22.2 1010.1, CE
Up to 80%
113°F
15°C to 30°C
7.08 in.
18 cm
3.14 in.
5.51 in.
1000 V
500 mA
Wide-Mini Vertical Electrophoresis Unit, Basic
Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, spacer-mate alignment template, wonder wedge plate separation tool and gel seal, bubble level
45°C
12.59 in.
32 cm
11.41 in.
29 cm
14 cm
50 w