Learn More
Macherey-Nagel™ Sea Sand
Use to prepare columns for use in low-pressure flash chromatography. Macherey-Nagel™ Sea Sand is acid-washed and calcined, and is available in 1kg bottle.
Brand:  Macherey-Nagel™ 727423
Product Code. 12706561
  • 48.95 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
When packing a column for use in low-pressure flash chromatography, users must ensure that the adsorbent cannot be flushed from the column and that the adsorbent does not become suspended in the eluent. Sea sand is placed above and below the silica to ensure a stable gel bed.
  • A pad of glass fiber wadding is placed in front of the tap and a layer of sea sand (about 0.5–1.0cm) is filled in above it
  • Then the dry adsorbent is packed into the column and is settled down until it forms a stable gel bed
  • The silica bed is covered with another layer of sea sand (about 0.5–1.0cm) to avoid suspension of the silica surface
Specifications
Specifications
Sea Sand, acid-washed and calcined
Sea Sand
Low Pressure Flash Chromatography
Flash Chromatography Glass Columns