Learn More
DWK Life Sciences Sealing ring, for polished spherical joints (silicone + FEP)
For sealing DURAN™ ball joints without ground joints with high demands on chemical resistance.
Brand: DWK Life Sciences 286231301
This item is not returnable.
View return policy
Description
Non-sanded DURAN™ spherical ground joints only seal via the sealing ring. FEP-coated silicone rings are used where high chemical resistance is required.
- Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
- Silicone and FEP sealing ring
Specifications
S 13 NS | |
Silicone, FEP | |
10/Pk. |
Not Autoclavable | |
Sealing of non-sanded spherical ground joints |