Learn More
DWK Life Sciences Sealing ring, for polished spherical joints (silicone + FEP)
For sealing DURAN™ ball joints without ground joints with high demands on chemical resistance.
Brand:  DWK Life Sciences 286231901
 View more versions of this product
Product Code. 15867081
  • 237.00 € / Pack of 10
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Non-sanded DURAN™ spherical ground joints only seal via the sealing ring. FEP-coated silicone rings are used where high chemical resistance is required.
  • Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
  • Silicone and FEP sealing ring
TRUSTED_SUSTAINABILITY
Specifications
Specifications
19 NS
Silicone, FEP
10/Pk.
Not Autoclavable
Sealing of non-sanded spherical ground joints