missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Seamless Cellulose Dialysis Tubing

Retains materials with 12,000 and higher molecular weight.

19.00 €

Specifications

Product Type Dialysis Tubing
Material Seamless Cellulose
Formulation Glycerin, water and 1% sulfur
Quantity Each
For Use With (Application) Useful in analytical chemistry, biology, and pharmaceutical fields
View More Specs

Products 1
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
17780245
View Documents
Fisher Science Education™
S25645A
20.4 mm 3 m
19.00 €
1 roll
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Especially useful in analytical chemistry, biology, and pharmaceutical fields
  • Highly permeable to water and low molecular weight compounds
  • Contains glycerin, water and 1% sulfur
  • Must be stored in airtight container or rehumidified before use
  • Pore diameter: 4.8nm

Specifications

Specifications

Dialysis Tubing
Glycerin, water and 1% sulfur
Useful in analytical chemistry, biology, and pharmaceutical fields
Seamless Cellulose
Each
Videos
SDS
Documents

Documents

Product Certifications