Learn More
sec-Butylamine, 99%, Thermo Scientific Chemicals
18.50 € - 137.00 €
Chemical Identifiers
CAS | 13952-84-6 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.14 |
MDL Number | MFCD00008094 |
InChI Key | BHRZNVHARXXAHW-UHFFFAOYNA-N |
Synonym | sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote |
PubChem CID | 24874 |
ChEBI | CHEBI:74526 |
IUPAC Name | butan-2-amine |
SMILES | CCC(C)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10695712
|
Thermo Scientific Acros
107810050 |
5 mL | Glass bottle |
18.50 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10529823
|
Thermo Scientific Acros
107810250 |
25 mL | Glass bottle |
22.60 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10792875
|
Thermo Scientific Acros
107811000 |
100 mL | Glass bottle |
26.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10026193
|
Thermo Scientific Acros
107815000 |
500 mL | Glass bottle |
41.30 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15179163
|
Thermo Scientific Acros
107810025 |
2.5 L | Glass bottle |
137.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13952-84-6 | |
73.14 | |
BHRZNVHARXXAHW-UHFFFAOYNA-N | |
24874 | |
butan-2-amine |
C4H11N | |
MFCD00008094 | |
sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote | |
CHEBI:74526 | |
CCC(C)N |
Specifications
13952-84-6 | |
Colorless to Yellow | |
63.0°C | |
Authentic | |
C4H11N | |
1.3920 to 1.3960 | |
04,16 | |
0.72 | |
Solubility in water: miscible. Other solubilities: freely miscible with most common organic solvents | |
CCC(C)N | |
73.14 | |
CHEBI:74526 | |
73.13 | |
Liquid |
-104.0°C | |
0.7200g/mL | |
−19°C | |
98.5% min. (GC) | |
C2H5CH(NH2)CH3 | |
MFCD00008094 | |
15,1546 | |
sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote | |
BHRZNVHARXXAHW-UHFFFAOYNA-N | |
butan-2-amine | |
24874 | |
0.5 mPa.s (20°C) | |
99% | |
sec-Butylamine |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-732-7
RUO – Research Use Only