missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15455405
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent
TRUSTED_SUSTAINABILITY

Specifications

CAS 171860-66-5
Density 0.968
Molecular Formula C4H9BrZn
MDL Number MFCD00671990
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 2-butylzinc bromide, bromo sec-butyl zinc, zinc, bromo 1-methylpropyl, sec-butylzinc bromide, 0.50 m in thf, 2-butylzinc bromide 0.5 m in tetrahydrofuran, sec-butylzinc bromide solution, 0.5 m in thf, sec-butylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key DATGUFISTUEOBS-UHFFFAOYSA-M
SMILES CC[CH-]C.[Zn+]Br
IUPAC Name bromozinc(1+);butane
Molecular Weight (g/mol) 202.4
PubChem CID 4101435
Formula Weight 202.4
Chemical Name or Material sec-Butylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.