missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art Secador™ 5.0 Adjustable Auto-Desiccator Cabinet, Electric Non-Desiccant

Product Code. 17805402 Shop All SP Scienceware Products
Click to view available options
:
Hinged Cabinet
This item is not returnable. View return policy

Product Code. 17805402

Brand: Bel-Art H420571004

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This cabinet provides dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament. This cabinet is an adjustable electric model that can be programed to your desired humidity level.

  • High-density foam gaskets to keep ambient humidity out and dry air inside
  • Large clear polycarbonate panels are durable and allow for easy viewing of internal contents
  • LCD shows the internal temperature and humidity level
  • Includes 3 adjustable and removable shelves
  • Electric models keeps internal humidity at 20 to 40%

Specifications

Autoclavable Not Autoclavable
Color White
Cabinet Style Front-loading
Material Polycarbonate
Door Style Hinged
No. of Shelves 3
Shelf Positions Adjustable
Dimensions (D x W x H) Interior 34 x 28 x 50 cm
Width (English) 11.8 in.
Width (Metric) 30 cm
Height (English) 25 in.
Height (Metric) 63.5 cm
Depth (English) 13 in.
Depth (Metric) 34 cm
Voltage 100/120 V or 200/240 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.