Learn More
Sartorius™ Secura™ Laboratory Balance with Internal Calibration
Laboratory balance with internal calibration and continuous real-time level monitoring.
Brand:  Sartorius™ SECURA1103-1CEU
6251.00 EUR valid until 2024-12-31
Use promo code "21987" to get your promotional price.
 View more versions of this product
Product Code. 17721818
  • 6580.00 € / Each
Explore more special offers
Description
Description
  • Built-in protection systems for complete reliability and regulatory compliance
  • Provides highly accurate weighing results and operating convenience
  • Real-time guidance prompts for leveling, automatic internal adjustment, and 100% traceable, clear documentation with sample and batch identifiers make your lab work more efficient
  • Real-time alarm messaging and interactive user guidance for leveling to prevent inaccurate results due to an unleveled balance
  • SQmin for automatic detection and active monitoring to ensure you are within acceptable USP operating range
  • Fully automatic internal adjustment function isoCAL for the highest possible accuracy at all times
  • Cal Audit Trail for easy, traceable documentation of all calibration and adjustment data
  • Safe and clear documentation by sample and batch identifiers for keeping perfect track of weighing data
  • GLP- and GMP-compliant printouts
  • Standard password security features for protection against tampering and unauthorized access
  • Integrated PC direct function and Plug and Work connectivity to a Sartorius™ printer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006
Color Touchscreen
Universal Power Adapter, In-use Cover, Application Guide, Dust Cover
Electronic Toploading Balances
100/240 V, 50/60 Hz
1100 g
1 mg
Stainless Steel
0.9 s