Learn More
Sartorius™ Secura™ Precision Laboratory Balance
Laboratory balance with internal calibration and continuous real-time level monitoring.
Brand:  Sartorius™ SECURA3101-1S
Product Code. 17741818
  • 2520.00 € / Each
Explore more special offers
Description
Description
Advanced Level Monitoring and Security Secura™ Precision Balances comes with a host of advanced features to ensure the most accurate and precise results possible.
  • Equipped with an advanced internal monitoring system that automatically calibrates your balance according to your password-protected menu settings
  • Automatic calibration and adjustment system monitors and reacts to temperature changes, eliminating result errors and saving time
  • Fully compliant data transfer capabilities and SQmin function to ensure reliable USP compliance
  • Choose from three levels of security options to customize to the needs of your laboratory
  • Ships with a series of integrated applications that significantly expand the potential of your balance from day one
  • Included applications allow researchers to carry out advanced weight measurements of mixtures, measure the density of both liquid and solid samples, make complex multi-step conversions and create calibration and weight statistical measurement reports
  • Balances arrive with an easy-to-navigate SmartTouch user display for easy menu navigation and application selection right out of the box
  • One-Touch draft shield is designed to minimize ambient effects on measurements while protecting the integrity of your samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006
Color Touchscreen
Stainless Steel
0.9 s
3100 g
100 mg
Electronic Toploading Balances
100/240 V, 50/60 Hz