Learn More
Sartorius Secura™ Topload Weighing Balances: Promo
GREENER_CHOICE
Designed for work in regulated environments for high quality low risk results. Sartorius Secura™ ensures the quality of the measurement results, compliance with regulatory requirements and work flow efficiency improvement.
Brand:  Sartorius SECURA6102-1S
Product Code. 15440573
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Weighing Range: 6100g; Readability; 10mg; Repeatability: 10mg; Linearity: 20mg
  • Promo offer expires 12-31-2016
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6.1 kg
10 mg
Universal power adapter, USB data cable, in-use co
6102
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products