Learn More
3M™ Secure Click™ Half Mask Reusable Respirator
Designed with smart and intuitive features, the Secure Click™ Half Mask Reusable Respirator is simple, comfortable and reliable. Available in three sizes: small, medium, and large, these respirators all come with an optional speaking diaphragm.
Brand:  3M™ 7100171980
41.04 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16446370
  • 60.60 € / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Greater breathability and comfort provided by the world's first quad flow cartridge system.
  • Push button seal check helps bolster confidence that the respirator is properly sealed to the face.
  • Speaking diaphragm is designed to help provide easier communication while you work.
  • Confidence that filters and cartridges are installed properly provided by all new Secure Click™ connection.
  • Low profile half mask design offers a wide field of view and compatibility with certain welding and grinding shields.
  • Over-moulded design for easy assembly/disassembly.
  • Advanced silicone material for increased comfort and greater durability.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AS/NZS 1716:2012
Dark Gray
Half Facepiece Respirator
Speaking Diaphragm
10/Cs.
Secure Click™
Silicone, Nylon
Medium
HF-802 SD
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Secure Click™ Half Mask Reusable Respirator > Half Mask Reusable Respirator; Size: Medium; Includes: Speaking Diaphragm