Learn More
3M™ Secure Click™ Reusable Full Face Respirator
The Secure Click™ Reusable Full Face Mask FF-800 Series helps protect against particulates and a wide variety of gases and vapors when used with approved Secure Click™ filters.
Brand:  3M™ 7100262628
285.93 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17956640
  • 439.00 € / Each
Explore more special offers
Description
Description
  • The quick, easy, one-touch positive pressure push button seal check helps provide improved confidence in respirator fit
  • Two dual-flow gas and vapor filters on each respirator combine to create the world's first quad flow gas and vapor filter system with four airflow paths providing easy breathing and comfort
  • Large lens for excellent field of vision, and treated with Scotchgard™ Protector for improved paint and stain resistance
  • Six-strap harness helps provide a secure comfortable fit
  • Speaking diaphragm is designed to help provide easy communication
  • Recommended applications include assembly, chemical clean-up, chemical handling, chipping, chiseling, cleaning, furnace operations, grinding, machining, masonry, painting, pouring/casting, sanding and sawing.
  • Available in three sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Secure Click™ Filters
Medium