missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ SecureFit™ X5000 Safety Helmet, Vented, 1000 V, CE
3M™ SecureFit™ Safety Helmet has six different settings to adjust position and a 4-point chin strap selector system. Vented and brimless safety hat with 3M™ UVicator™ Sensor that indicates damaging levels of UV light.
511.00 €
Specifications
For Use With (Application) | Construction, Forestry, Oil, and Gas |
---|---|
Material | ABS |
Product Type | Safety Helmet |
Suspension Type | Pressure Diffusion 6 Point Ratchet |
Ventilated | Ventilated |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17355323
|
3M™
7100175101 |
White |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17365323
|
3M™
7100175511 |
Yellow |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17375323
|
3M™
7100175512 |
Blue |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17385323
|
3M™
7100175533 |
Green |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17395323
|
3M™
7100175534 |
Red |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17305333
|
3M™
7100175535 |
Orange |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17315333
|
3M™
7100175536 |
Black |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17325333
|
3M™
7100175537 |
Hi-vis Green |
511.00 €
Pack of 4 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
Description
- Secure, comfortable head protection for working at heights and other hazardous industrial jobs
- Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
- 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
- 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
- Inspired by climbing helmets with a brimless design to give you a clear upward view
- Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
- Certified to either EN12492 or EN397 (depending on chin strap configuration) and EN50365 1000 Vac standard
- Approved for lateral deformation (LD)
- UV-stabilized ABS
Specifications
Construction, Forestry, Oil, and Gas | |
Safety Helmet | |
Ventilated |
ABS | |
Pressure Diffusion 6 Point Ratchet |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title