missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ SecureFit™ X5000 Safety Helmet, Vented, Reflective, CE
3M™ SecureFit™ Safety Helmet has six different settings to adjust position and a 4-point chin strap selector system. Vented and brimless safety helmet with 3M™ UVicator™ Sensor that indicates damaging levels of UV light.
570.00 €
Specifications
For Use With (Application) | Construction, Forestry, Oil, and Gas |
---|---|
Material | ABS |
Product Type | Safety Helmet |
Suspension Type | Pressure Diffusion 6 Point Ratchet |
Ventilated | Ventilated |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17375313
|
3M™
7100175093 |
White |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17385313
|
3M™
7100175094 |
Yellow |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17395313
|
3M™
7100175095 |
Blue |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17305323
|
3M™
7100175096 |
Green |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17315323
|
3M™
7100175097 |
Red |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17325323
|
3M™
7100175098 |
Orange |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17335323
|
3M™
7100175099 |
Black |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
17345323
|
3M™
7100175100 |
Hi-vis Green |
570.00 €
Pack of 4 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
Description
- Secure, comfortable head protection for working at heights and other hazardous industrial jobs
- Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
- 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
- 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
- Magnetic buckle clip as standard
- Inspired by climbing helmets with a brimless design to give you a clear upward view
- Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
- Certified to either EN12492 or EN397 (depending on chin strap configuration)
- Approved for lateral deformation (LD)
- UV-stabilized ABS
Specifications
Construction, Forestry, Oil, and Gas | |
Safety Helmet | |
Ventilated |
ABS | |
Pressure Diffusion 6 Point Ratchet |