Learn More
WLD Tec™ Security Adapter
Brand:  WLD Tec™ 6.000.800
Product Code. 10487151
  • 172.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Order Info
ProductTypeSpecs
Specifications
Specifications
Gas Adapter
0.5 m
DVGW Tubing (0.5 m) for Cartridges C 206, 190 g from Camping Gaz, Integrated Pressure Regulator 50 mbar, Shut-Off Valve