missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Imhoff Cone Stand
Securely hold two Imhoff cones with this durable cone stand. Kartell™ Imhoff Cone Stand provides a dependable place to observe settleable solids in surface and saline waters or sewage.
Brand: Kartell™ 0105600
This item is not returnable.
View return policy
Description
Securely hold two Imhoff cones with this durable cone stand. Kartell™ Imhoff Cone Stand provides a dependable place to observe settleable solids in surface and saline waters or sewage.
- Holds two Imhoff cones
- Constructed of PMMA
- Height of 294mm
Order Info
LengthMetric
Specifications
PMMA | |
Sedimentation Imhoff Cone Stand | |
150 mm | |
294 mm |
2 | |
150 x 300 x 294 mm | |
300 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction