Learn More
Cytiva Select-a-Dye mono-Reactive Dye Packs
Enables a variety of labeling strategies using NHS ester, maleimide, or hydrazide to be investigated
Brand:  Cytiva PA15123
Product Code. 11832460
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Available as either Cy3 or Cy5
  • Example protocols included with each dye pack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Steady state Fluorescence, FP and FRET
Cy5
3 x 0.5 mg
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1