Learn More
3M™ Petrifilm™ Select E. Coliplate
Encompass
Specific for E. coli, only E. coli colonies are visible which numerate within 24 hours
POA
Specifications
Detects E. coli
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17223784
Encompass
View Documents
3M™ Petrifilm™
7100126813
500 Pack
POA
Pack of 500
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17233784
Encompass
View Documents
3M™ Petrifilm™
7100126814
25 Pack
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Specific test for E. coli, only E. coli colonies are visible
  • E. coli enumerates in just 24 hours
  • Contains a ß-glucuronidase indicator
Specifications
Specifications
E. coli
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 18 Months
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Petrifilm™ Select E. Coliplate