missing translation for 'onlineSavingsMsg'
Learn More

Selenic acid, 40% aq. soln., Thermo Scientific Chemicals

Product Code. 11343497
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-08-6
MDL Number MFCD00012191

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 g
Molecular Formula H10N2O4S
Linear Formula H2SeO4
UN Number UN2922
Merck Index 14,8429
Solubility Information Miscible with sulfuric acid. Immiscible with ammonia.
InChI Key ASPXBYAQZVXSNS-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].O.[O-]S(=O)[O-]
IUPAC Name diazanium;sulfite;hydrate
Molecular Weight (g/mol) 134.15
PubChem CID 21701053
Formula Weight 144.97 (Anhydrous)
Concentration or Composition (by Analyte or Components) 40% aq. soln.
Density 1.41 g/mL
Chemical Name or Material Selenic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'