missing translation for 'onlineSavingsMsg'
Learn More
Selenic acid, 40% aq. soln., Thermo Scientific Chemicals Product Code.: 11353497

Selenic acid, 40% aq. soln., Thermo Scientific Chemicals

Product Code. 11353497
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11353497

Brand: Thermo Scientific Alfa Aesar 018851.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selenic acid is used as an oxidizing agent. Its derivative sodium selenate is used in the preparation of glass and animal feeds. It acts as an analytical reagent and also used for the preparation of other selenium salts such as gold(III) selenate. Further, it reacts with fluorosulfuric acid to get selenoyl fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-08-6
MDL Number MFCD00012191
Synonym selenic vi acid, h2seo4, unii-hv0y51nc4j, hv0y51nc4j, hsdb 675, dihydroxidodioxidoselenium, h2so4e, aq. soln., seo2 oh 2
ChEBI CHEBI:18170

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 g
Molecular Formula K2O4Se
Linear Formula H2SeO4
UN Number UN2922
Merck Index 14,8429
Solubility Information Miscible with sulfuric acid. Immiscible with ammonia.
InChI Key YAZJAPBTUDGMKO-UHFFFAOYSA-L
SMILES O[Se](=O)(=O)O
IUPAC Name selenic acid
Molecular Weight (g/mol) 221.16
PubChem CID 1089
Formula Weight 144.97 (Anhydrous)
Concentration or Composition (by Analyte or Components) 40% aq. soln.
Density 1.41 g/mL
Chemical Name or Material Selenic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenic acid, 40% aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.