missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selenious acid, 98%, Thermo Scientific Chemicals
69.35 € - 621.09 €
Chemical Identifiers
CAS | 7783-00-8 |
---|---|
Molecular Formula | H2O3Se |
Molecular Weight (g/mol) | 128.98 |
MDL Number | MFCD00011331 |
InChI Key | MCAHWIHFGHIESP-UHFFFAOYSA-N |
Synonym | selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution |
PubChem CID | 1091 |
ChEBI | CHEBI:26642 |
IUPAC Name | selenous acid |
SMILES | O[Se](O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10002480
|
Thermo Scientific Acros
198870500 |
50 g | Glass bottle |
69.35 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276780
|
Thermo Scientific Acros
198872500 |
250 g | Glass bottle |
225.12 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904111
|
Thermo Scientific Acros
198870010 |
1 kg | Glass bottle |
621.09 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-00-8 | |
128.98 | |
MCAHWIHFGHIESP-UHFFFAOYSA-N | |
1091 | |
selenous acid |
H2O3Se | |
MFCD00011331 | |
selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution | |
CHEBI:26642 | |
O[Se](O)=O |
Specifications
Pink or White | |
70.0°C | |
H2O3Se | |
MFCD00011331 | |
158567 | |
Solubility in water: 167g/100mL (20°C). Other solubilities: very soluble in hot water and alcohol,insoluble in ammonia | |
O[Se](O)=O | |
128.98 | |
CHEBI:26642 | |
98% |
7783-00-8 | |
97.5% min. (Iodometry) | |
Crystalline Powder, Crystals and/or Chunks | |
1992 | |
selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution | |
MCAHWIHFGHIESP-UHFFFAOYSA-N | |
selenous acid | |
1091 | |
128.97 | |
Selenious acid |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-974-7
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title