Learn More
Selenium, 99+%, powder, Thermo Scientific Chemicals
111.00 € - 348.00 €
Chemical Identifiers
CAS | 7782-49-2 |
---|---|
Molecular Formula | Se |
Molecular Weight (g/mol) | 78.97 |
MDL Number | MFCD00134090 MFCD00011224 |
InChI Key | BUGBHKTXTAQXES-UHFFFAOYSA-N |
Synonym | selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray |
PubChem CID | 6326970 |
ChEBI | CHEBI:27568 |
IUPAC Name | selenium |
SMILES | [Se] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10289700
|
Thermo Scientific Acros
419271000 |
100 g | Glass bottle |
111.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10764014
|
Thermo Scientific Acros
419275000 |
500 g | Glass bottle |
348.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-49-2 | |
78.97 | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
6326970 | |
selenium |
Se | |
MFCD00134090 MFCD00011224 | |
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray | |
CHEBI:27568 | |
[Se] |
Specifications
685°C | |
7782-49-2 | |
Powder | |
Se | |
MFCD00134090 MFCD00011224 | |
15, 8568 | |
Solubility in water: insoluble. Other solubilities: soluble in concentrated acid,slightly soluble in diehtyl ether,almost insoluble in carbon disulfide,insoluble in ethanol | |
[Se] | |
78.97 | |
CHEBI:27568 | |
≥99% | |
4.81 | |
Selenium, <325 mesh |
217°C | |
Cream-White to White | |
99% min | |
Se | |
01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281 | |
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
selenium | |
6326970 | |
78.96 | |
0.5% max | |
4.8100g/mL |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
May cause long lasting harmful effects to aquatic life.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-957-4
- Selenium
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H413-May cause long lasting harmful effects to aquatic life.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only