missing translation for 'onlineSavingsMsg'
Learn More

Selenium(I) chloride, 99%

Product Code. p-7051006
Click to view available options
Quantity:
25 g
2 x 25 g
Unit Size:
25g
50g
This item is not returnable. View return policy

Product Code. 11323758

Brand: Thermo Scientific Alfa Aesar 040239.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-68-0 | Cl2Se2 | 228.84 g/mol

Selenium monochloride is used in electronics, instrumentation and other industrial purposes. It is used as a reagent for the synthesis of Se-containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-68-0
Molecular Formula Cl2Se2
Molecular Weight (g/mol) 228.84
MDL Number MFCD00011226
InChI Key VIEXQFHKRAHTQS-UHFFFAOYSA-N
Synonym diselenium dichloride, selenium monochloride, selenium chloride se2cl2, unii-2a5292fc4p, selenium i chloride, dichlorodiselenane, dichloro perselenide, se2cl2, selenium i chloride, ampuled under argon trace metals basis 25g
PubChem CID 66206
IUPAC Name chloroselanyl selenohypochlorite
SMILES Cl[Se][Se]Cl

Specifications

Boiling Point 127°C (decomposition)
Melting Point -85°C
Physical Form Liquid
Quantity 25 g
UN Number UN2922
Merck Index 14,8432
Solubility Information Decomposes in water; Soluble in chloroform, benzene, carbon tetrachloride, carbon disulfide
Formula Weight 228.83
Percent Purity 99%
Odor Acidic
Sensitivity Moisture sensitive
Density 2.77 g/mL
Chemical Name or Material Selenium(I) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.