missing translation for 'onlineSavingsMsg'
Learn More
Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals Product Code.: 11470251

Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals

Product Code. 11470251
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11470251

Brand: Thermo Scientific Alfa Aesar 040240.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-65-3
Molecular Formula Br4Se
Molecular Weight (g/mol) 398.587
MDL Number MFCD00049766
InChI Key VTQZBGAODFEJOW-UHFFFAOYSA-N
Synonym selenium tetrabromide, selenium iv bromide, selenium bromide sebr4 , t-4, unii-8o54yxr180, tetrabromo-, e4-selane, sebr4, tetrabromo-$l^ 4-selane, tetrabromo-??-selane, selenium iv bromide 5g
PubChem CID 82246
SMILES [Se](Br)(Br)(Br)Br

Specifications

Melting Point 70°C to 80°C (decomposition)
Physical Form Powder
Quantity 5 g
UN Number UN2923
Merck Index 14,8437
Solubility Information Decomposes in water; Soluble in carbon disulfide, chloroform, ethyl bromide
Formula Weight 398.6
Percent Purity ≥99%
Sensitivity Moisture sensitive
Chemical Name or Material Selenium(IV) bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium(IV) bromide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.