missing translation for 'onlineSavingsMsg'
Learn More
Selenium(IV) oxide, 99.4% (metals basis), Thermo Scientific Chemicals Product Code.: 11311739

Selenium(IV) oxide, 99.4% (metals basis), Thermo Scientific Chemicals

Product Code. 11311739
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11311739

Brand: Thermo Scientific Alfa Aesar 012358.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the manufacture of other selenium compounds. Selenium dioxide is an oxidizing agent mostly used in the allylic oxidation of alkenes to produce allylic alcohols. It is used in creating colorless glass and toner in photographic developing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-08-4
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
SMILES O=[Se]=O

Specifications

Melting Point 340°C to 350° (sublimation)
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.4% (Metals Basis)
Solubility Information Partially soluble in water,methanol,ethanol,and acetone. Soluble in concentrated sulfuric acid.
IUPAC Name selenium dioxide
Formula Weight 110.96
Odor Odorless
Density 3.95 g/cm3 at 20°C
Chemical Name or Material Selenium(IV) oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Selenium (IV) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Selenium dioxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium(IV) oxide, 99.4% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.