missing translation for 'onlineSavingsMsg'
Learn More
Selenium(IV) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10358620

Selenium(IV) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10358620
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10358620

Brand: Thermo Scientific Acros 193980100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-08-4
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
IUPAC Name selenium dioxide
SMILES O=[Se]=O

Specifications

Melting Point 315°C
Color White
Physical Form Fine Crystalline Powder
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula SeO2
Fieser 01,992; 02,360; 03,245; 04,422; 05,575; 06,509; 08,439; 09,409; 10,345; 11,96; 13,272; 14,177; 15,89; 17,312
Merck Index 15, 8571
Solubility Information Solubility in water: 38.4g/100mL (14°C). Other solubilities: 1.11 parts/100 parts acetic acid (13.9°C),10.16 parts/100 parts methanol (11.8°C),6.67 parts/100 parts 93% ethanol (14°C),4.35 parts/100 parts acetone (15.3°C)
Formula Weight 110.96
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Selenium(IV) oxide, 99.999%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Selenium(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium(IV) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.