missing translation for 'onlineSavingsMsg'
Learn More
Selenium(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11327007

Selenium(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11327007
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11327007

Brand: Thermo Scientific Alfa Aesar 010854.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In organic reactions, selenium dioxide is often used as an oxidizing reagent employed for carrying out allylic oxidations, Riley oxidation, synthesis of diones, etc. It is used to impart a red color to glass. It finds use as a toner in photographic developing, and an active ingredient in some cold-blueing solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-08-4
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
IUPAC Name selenium dioxide
SMILES O=[Se]=O

Specifications

Melting Point 340°C to 350°C (sublimation)
Physical Form Lump/Powder
Quantity 5 g
UN Number UN3283
Merck Index 14,8434
Solubility Information Soluble in water,benzene,ethanol and methanol. Sprangly soluble in acetone and acetic acid.
Formula Weight 110.96
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Assay (metals basis)
Chemical Name or Material Selenium(IV) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.