missing translation for 'onlineSavingsMsg'
Learn More

Selenium shot, amorphous, 1-6mm (0.04-0.24in), Puratronic, 99.999+% (metals basis), Thermo Scientific Chemicals

78.99 € - 299.49 €

Chemical Identifiers

CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366507
View Documents
Thermo Scientific Alfa Aesar
000290.18
50 g
78.99 €
50g
Please to purchase this item. Need a web account? Register with us today!
11356507
View Documents
Thermo Scientific Alfa Aesar
000290.30
250 g
299.49 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-49-2
78.97
BUGBHKTXTAQXES-UHFFFAOYSA-N
6326970
selenium
Se
MFCD00134090 MFCD00011224
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
CHEBI:27568
[Se]

Specifications

7782-49-2
>99.999% (Metals basis)
MFCD00134090 MFCD00011224
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
6326970
Odorless
Selenium
Shot
Se
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
[Se]
78.97
CHEBI:27568
Various colors shot
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 231-957-4

missing translation for 'rtecsNumber' : VS7700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Selenium shot

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 4

Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.