missing translation for 'onlineSavingsMsg'
Learn More

Selenium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

Product Code. 10458600
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

MDL Number 134090

Specifications

Packaging HDPE plastic bottle
Quantity 500 mL
Color Colorless
pH 1
Physical Form Liquid
compliance-icons
Product Identifier
  • Selenium Standard Metal Solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'