missing translation for 'onlineSavingsMsg'
Learn More

Selenium sulfide, 94%, Thermo Scientific Chemicals

56.48 € - 550.74 €

Chemical Identifiers

CAS 7488-56-4
Molecular Formula S2Se
Molecular Weight (g/mol) 143.09
MDL Number MFCD00011227
InChI Key JNMWHTHYDQTDQZ-UHFFFAOYSA-N
Synonym selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
PubChem CID 24087
IUPAC Name thione
SMILES S=[Se]=S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11390208
SDS View Documents
Thermo Scientific Alfa Aesar
042943.14
25 g
56.48 €
25g
Please to purchase this item. Need a web account? Register with us today!
11380208
SDS View Documents
Thermo Scientific Alfa Aesar
042943.22
100 g
157.79 €
100g
Please to purchase this item. Need a web account? Register with us today!
11370208
SDS View Documents
Thermo Scientific Alfa Aesar
042943.36
500 g
550.74 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7488-56-4
143.09
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
24087
S=[Se]=S
S2Se
MFCD00011227
selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
thione

Specifications

(decomposition)
7488-56-4
Powder
S2Se
selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
thione
24087
Selenium sulfide
<100°C
Red to Brown
94%
MFCD00011227
Insoluble in water,in dilute HCl
S=[Se]=S
143.09
143.09
Videos
Safety and Handling

Safety and Handling

P262-P264b-P270-P280-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 231-303-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Selenium sulphide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : Selenium sulphide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.