missing translation for 'onlineSavingsMsg'
Learn More

Selenourea, 98+%, Thermo Scientific Chemicals

56.08 € - 233.16 €

Chemical Identifiers

CAS 630-10-4
Molecular Formula CH4N2Se
Molecular Weight (g/mol) 123.02
MDL Number MFCD00008065
InChI Key ZUCFIOXGLBMWHV-UHFFFAOYSA-N
Synonym selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
PubChem CID 6327594
IUPAC Name $l^{1}-selanylmethanimidamide
SMILES C(=N)(N)[Se]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10702261
View Documents
Thermo Scientific Acros
213170010
1 g Glass bottle
56.08 €
1g
Please to purchase this item. Need a web account? Register with us today!
10599813
View Documents
Thermo Scientific Acros
213170050
5 g Glass bottle
233.16 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

630-10-4
123.02
ZUCFIOXGLBMWHV-UHFFFAOYSA-N
6327594
C(=N)(N)[Se]
CH4N2Se
MFCD00008065
selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
$l^{1}-selanylmethanimidamide

Specifications

630-10-4
Gray to Pink
98% min
H2NC(=Se)NH2
selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
ZUCFIOXGLBMWHV-UHFFFAOYSA-N
$l^{1}-selanylmethanimidamide
6327594
98+%
Selenourea
214.0°C to 215.0°C
Authentic
CH4N2Se
MFCD00008065
Solubility in water: very soluble.
C(=N)(N)[Se]
123.02
123.02
Crystalline Powder or Needles
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 211-129-9

SDS
Documents

Documents

Certificates

RUO – Research Use Only