Learn More
Brady™ Self-laminating Aggressive Adhesive Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
High-adhesion Self-laminating Vinyl Wraps are for tough conditions, including high-humidity environments, small diameter wires and newer wire jacketing materials.
Brand:  Brady™ 174393
 View more versions of this product
Product Code. 17828944
  • 63.10 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dependable, long-term adhesion to small diameter wires
  • Will stay adhered in even the most humid environments
  • Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC
  • Features a white printable area and a clear overlaminating area that protects the print
  • Ideal for control panels, electrical panels, wire harnesses, datacom and wire and cable ID
  • Made of B-417 vinyl material
Specifications
Specifications
Not Autoclavable
M6-18-417
None
Matte
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
250 Labels
-40°F to +158°F
0.004 in.
1 x 0.75 in.
0.75 in.
0.75 in.
0.375 in.
9.53 mm
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Permanent Acrylic
Clear, White
Die-cut
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling, Wire and Cable Labeling
No
Rectangle
-40°C to +70°C
0.107 mm
1 in.
19.05 mm
19.05 mm
25.4 mm