Learn More
Brady™ Self-laminating Cryogenic Polyester Laboratory Labels for M6/M7 Printers
Ideal for lab applications, these ultra-thin, self-laminating labels protect text on vials, tubes, straws and slides. Use in liquid nitrogen and autoclave applications.
Brand:  Brady™ 174149
 View more versions of this product
Product Code. 17839134
  • 54.30 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Performs well in liquid nitrogen and autoclave environments; temperature ranges from -196°C to 130°C (-320°F to 266°F)
  • Resists exposure to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% clorox solution
  • White printable zone on clear polyester with black printing
  • RoHS compliant to 2011/65/EU (Pass 2015/863)
  • Made of B-461 polyester material
Specifications
Specifications
Autoclavable
Permanent Acrylic
White
Continuous
Single-side Printable
Labels
Component and Equipment Labeling, General Identification, Laboratory Labeling
No
Rectangle
-196°C to +110°C
0.048 mm
50 ft.
0.375 in.
M6C-375-461
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 ft.
-321°F to +230°F
0.0019 in.
50 ft. x 0.375 in.
15.24 m
9.53 mm