Learn More
Brady™ Self-laminating Gray Vinyl Wrap Around Wire and Cable Label Tape for M6/M7 Printers
Self-laminating vinyl material is ideal for wire and cable identification. Designed with a clear protective layer to cover the printed text, B-427 material provides superb protection from abrasion, dirt, oils, solvents, water and excess handling.
44.80 € - 79.80 €
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Adhesive Type Permanent Acrylic
Coating None
Color Gray, Clear
View More Specs
Products 10
Product Code Brand Dimensions (LxW) Height (Metric) Shape Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Height (Metric) Shape Price Quantity & Availability  
17811549
View Documents
Brady™
173738
1 x 1 in. 25.4 mm Square
58.50 €
Pack of 250
17851549
View Documents
Brady™
173784
1.5 x 0.25 in. 38.1 mm Rectangle
73.25 €
Pack of 750
17831679
View Documents
Brady™
173797
1.5 x 0.5 in. 38.1 mm Rectangle
79.80 €
Pack of 500
17801579
View Documents
Brady™
173811
1.5 x 0.75 in. 38.1 mm Rectangle
56.70 €
Pack of 250
17851499
View Documents
Brady™
173823
1.5 x 1 in. 38.1 mm Rectangle
75.25 €
Pack of 250
17881439
View Documents
Brady™
173835
1.5 x 1.5 in. 38.1 mm Square
77.35 €
Pack of 250
17861509
View Documents
Brady™
173751
2.5 x 1 in. 63.5 mm Rectangle
44.80 €
Pack of 100
17881409
View Documents
Brady™
173762
4 x 1 in. 101.6 mm Rectangle
53.60 €
Pack of 100
17841559
View Documents
Brady™
173847
4 x 1.5 in. 101.6 mm Rectangle
72.55 €
Pack of 100
17871399
View Documents
Brady™
173855
6 x 1.5 in. 152.4 mm Rectangle
60.10 €
Pack of 50
Description
Description
  • Provided in a self-laminated format with a gray printable zone and translucent overlaminating area to protect text
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • UL approved
  • Service temperature of -40°C to +70°C (-40°F to +158°F)
  • Made of B-427 vinyl material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Gray, Clear
Die-cut
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
No
-70°C to +70°C
0.102 mm
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
None
Matte
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
-94°F to +158°F
0.004 in.