Learn More
Brady™ Self-Laminating Rotating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Identify your wires from any angle with Rotating Vinyl Wire Labels. Simply print your labels and then peel, fold and wrap – they’ll rotate freely for easy ID on even closed network connections.
Brand:  Brady™ 174168
 View more versions of this product
Product Code. 17818314
  • 53.85 € / Pack of 50
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rotates so you can identify your labels from any angle
  • Simply wrap and apply--there's no adhesive residue or label parts to cling to your wires
  • Spins freely without anchors so these labels won't damage your Cat 5 / 6 and fiber optic cables
  • Made of B-427 vinyl
Specifications
Specifications
Not Autoclavable
M6-207-427-RO
None
Matte
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 Labels
-94°F to +158°F
0.004 in.
2.75 x 0.5 in.
0.5 in.
0.5 in.
1.1 in.
27.94 mm
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Permanent Acrylic
Clear, White
Die-cut
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
No
Rectangle
-70°C to +70°C
0.102 mm
2.75 in.
12.7 mm
12.7 mm
69.85 mm