Learn More
Brady™ Self-laminating Vinyl MiniMark™ Wire and Cable Marker
This Self-laminating Vinyl MiniMark™ Wire and Cable Marker is made of conformable material designed to adhere to highly curved surfaces, wires and cables.
Brand:  Brady™ 710114
 View more versions of this product
Product Code. 11701825
  • 417.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features a white printable area with clear ''tail'' that wraps around marker.
  • The clear tails serves as over-laminate to protect the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling.
  • This material is UL recognised with its respective ribbon.
  • See the Recommended Ribbon Series for the appropriate ribbon to use.
Specifications
Specifications
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognised
MiniMark Self-laminating Wire and Cable Markers
White, Transparent
Label
Thermal Transfer
Data and telecommunications labelling, Electrical labelling, Laboratory labelling, Wire and cable labelling
-70°C
2780 Pk.
-94°F to +158°F
Tensile Strength and Elongation 12 lb./in. (210 N/100 mm), 152%, 11 lb./in. (193 N/100 mm), 195%
0.102 mm
1.25 in.
1 in.
0.5 in.
6.10 to 3.40 mm
MNK-BPT-311-427
Permanent Acrylic
Matte
Self-laminating Vinyl
Labels
MiniMark Printer
2780 Labels/Box
25.4 x 31.75 mm (W x H)
-70°C to +70°C
0.004 in.
31.75 x 25.40 mm
31.75 mm
25.4 mm
12.7 mm