Learn More
Brady™ Self-laminating Vinyl Wrap Around Labels
Mark cables and wires with the B-427 self-laminating vinyl label material, designed with a clear self-laminating protective layer. Print almost anywhere with a durable Brady™ portable label printer.
Brand:  Brady™ 170870
 View more versions of this product
Product Code. 17841789
  • 44.20 € / Pack of 140
Explore more special offers
Description
Description
  • Designed for tightly-curved surfaces such as wires and cables
  • Self-extinguishing and resistant to abrasion, solvents, oil, grease, weather and extreme temperatures
  • Made with a permanent acrylic adhesive that works great on smooth, rough, powder-coated and irregular surfaces
  • Made with a clear, non-printable overlaminate for increased protection
Specifications
Specifications
UL Recognized to UL 2238, RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Matte
Self-laminating Vinyl
Non-sterile
BMP51, M511
-94°F to +158°F
1.5 x 1.75 in.
1.75 in.
M5-134-427
Black on White/Transparent
Die-cut
Labels
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
140 Labels/Cartridge
0.004 in.
1.5 in.
4.5 cm