Learn More
Brady™ Self-laminating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Self-laminating vinyl material is ideal for wire and cable identification. Designed with a clear protective layer to cover the printed text, B-427 material provides superb protection from abrasion, dirt, oils, solvents, water and excess handling.
Brand:  Brady™ 174411
 View more versions of this product
Product Code. 17869104
  • 595.00 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • UL approved
  • Service temperature of -40°C to 70°C (-40°F to 158°F)
Specifications
Specifications
Not Autoclavable
BM-107-427
None
Matte
Vinyl
Thermal Transfer
Non-sterile
M610, M710, M611, BMP61, BMP71
1000 Labels
-94°F to +158°F
0.004 in.
1.5 x 2 in.
2 in.
2 in.
0.5 in.
12.7 mm
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Permanent Acrylic
Clear, White
Die-cut
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
No
Rectangle
-70°C to +70°C
0.102 mm
1.5 in.
50.8 mm
50.8 mm
38.1 mm