missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE

Self Seal Tyvek™ Steam Sterilization Pouches provide an excellent barrier from microbial contamination.

Brand:  Keystone Cleanroom Products BBMGTVKSS0810

1358.50 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 17747837

  • 1485.00 € / Pack of 500
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
  • The clear film side offers excellent clarity along with strength and durability
  • These bags feature a medical grade adhesive strip for convenient self-sealing
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates the need for the use and validation of a Heat Sealer
Specifications

Specifications

Autoclavable
Self-seal
Steam Sterilization
500/Pk.
Certificate of Conformance (COC)
White
HDPE
10 x 8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers