missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE

Self Seal Tyvek™ Steam Sterilization Pouches provide an excellent barrier from microbial contamination.

697.00 € - 4255.00 €

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Self-seal
Color White
For Use With (Application) Steam Sterilization
View More Specs

Products 44
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
17841701
View Documents
Keystone Cleanroom Products
00308TWBBSSSDSP02
3.5 x 8 in.
697.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17851701
View Documents
Keystone Cleanroom Products
00415TWBBSSSDSP02
4 x 15 in.
1205.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17861701
View Documents
Keystone Cleanroom Products
00510TWBBSSSDSP02
5 x 10 in.
882.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17707827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0206
6 x 2 in.
1300.00 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
17727827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0306
6 x 3 in.
1325.00 €
Pack of 1000
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17861711
View Documents
Keystone Cleanroom Products
01635TWBBSSSDSP02
6 x 35 in.
2340.00 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
17737827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0308
8 x 3 in.
744.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17747827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS03508
8 x 3.5 in.
759.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17787827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0608
8 x 6.125 in.
979.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17871701
View Documents
Keystone Cleanroom Products
00810TWBBSSSDSP02
8 x 10 in.
1300.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17881701
View Documents
Keystone Cleanroom Products
00815TWBBSSSDSP02
8 x 15 in.
1435.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17891701
View Documents
Keystone Cleanroom Products
00834TWBBSSSDSP02
8 x 34 in.
3055.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17717827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS02095
9 x 2.5 in.
1485.00 €
Pack of 1000
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17777827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0510
10 x 5 in.
1240.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17747837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0810
10 x 8 in.
1485.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17801711
View Documents
Keystone Cleanroom Products
01011TWBBSSSDSP02
10 x 11 in.
1670.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17811711
View Documents
Keystone Cleanroom Products
01020TWBBSSSDSP02
10 x 20 in.
2085.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17797827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0612
12 x 6 in.
1400.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17787837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1012
12 x 10 in.
2035.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17737837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0713
13 x 7 in.
1485.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17707847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1214
14 x 12 in.
2285.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17757827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0415
15 x 4 in.
1400.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17757837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0815
15 x 8.125 in.
1715.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17831711
View Documents
Keystone Cleanroom Products
01520TWBBSSSDSP02
15 x 20 in.
2920.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17841711
View Documents
Keystone Cleanroom Products
01626TWBBSSSDSP02
16 x 26 in.
3610.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17851711
View Documents
Keystone Cleanroom Products
01632TWBBSSSDSP02
16 x 32 in.
2340.00 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
17717847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1217
17 x 12 in.
2470.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17747847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1617
17 x 16 in.
3550.00 €
Pack of 500
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17767837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0818
18 x 8 in.
2195.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17797837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1020
20 x 10.125 in.
2405.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17737847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1520
20 x 15 in.
3435.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17767827
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0422
22 x 4 in.
1400.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17750368
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1822
22 x 18 in.
4210.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17727847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1424
24 x 14 in.
3980.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17757847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1626
26 x 16 in.
4255.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17777847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS2426
26 x 24 in.
2870.00 €
Pack of 250
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17707837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0628
28 x 6 in.
2285.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17760368
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS2430
30 x 24 in.
2985.00 €
Pack of 250
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17717837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0632
32 x 6 in.
3380.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17740368
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1632
32 x 16 in.
2710.00 €
Pack of 250
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17777837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0834
34 x 8 in.
3655.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17727837
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS0635
35 x 6 in.
3565.00 €
Pack of 500
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17787847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS2435
35 x 24 in.
3195.00 €
Pack of 250
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
17767847
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BBMGTVKSS1635
35.25 x 16 in.
3055.00 €
Pack of 250
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
  • The clear film side offers excellent clarity along with strength and durability
  • These bags feature a medical grade adhesive strip for convenient self-sealing
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates the need for the use and validation of a Heat Sealer
Specifications

Specifications

Autoclavable
Self-seal
Steam Sterilization
Certificate of Conformance (COC)
White
HDPE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.