missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Self-Sealing Sterilization Peel Pouch, Medical Paper/Film, With Steam Indicator
GREENER_CHOICE

Self-Sealing Steam Sterilization Pouches are designed for autoclave cycles

Brand:  Keystone Cleanroom Products PPSS10X14CF

 View more versions of this product

Product Code. 17921342

  • 279.93 € / Pack of 500

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Peels Open Cleanly
  • Triple Weld Factory seals
  • Convenient, time saving self-seal closure system
  • Strong, rip and tear resistant medical grade paper material
  • Good barrier characteristics prevent microbial penetration
  • Peels Open Cleanly
  • Sub-packed into sealed labeled poly bags with manufacturing lot number
  • Date of Manufacture on label for complete traceability
Specifications

Specifications

Self-sealing Sterilization Pouch
105 oz.
CoC
White
Steam Sterilization
Medical Grade Kraft Paper (Bleached)
47 to 57 μm
500/Cs.
60 gsm
10 in.
14 in.
Autoclavable
3.1 L
Self-adhesive
Single-use
Pharmaceutical parts
Non-sterile
Clear
121°C to 135°C
10 x 14 in.
25.4 cm
35.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers