missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Selinexor

Selective exportin 1 (XPO1/CRM1) inhibitor

211.00 € - 825.00 €

Specifications

Chemical Name or Material (2Z)-3-[3-[3,5-Bis(trifluoromethyl)phenyl]-1H-1,2,4-triazol-1-yl]-2-propenoic acid 2-(2-pyrazinyl)hydrazide
CAS 1393477-72-9
Target Antiviral Compounds
Molecular Formula C17H11F6N7O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18737176
View Documents
Tocris Bioscience™
7760/10
10 mg
211.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18736116
View Documents
Tocris Bioscience™
7760/50
50 mg
825.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Selinexor is an inhibitor of the exportin 1 (XPO1/CRM1) nuclear exporter. Shows selectivity for TNBC over ER-positive breast cancer cell lines (IC50 values are 11-550 nM and >1000 nM, respectively), and reduces growth of xenografts. Inhibits growth of thyroid cancer cell lines (IC50 = 150-500 nM), potentiates the anti-tumor effects of Olaparib (Cat. No. 7579) in ovarian cancer and breast cancer. Also a potent inhibitor of SARS-CoV-2 replication in vitro (EC50 = 10 nM) and downregulates the production of pro-inflammatory cytokines including TNF-α and IL-6.
Especificaciones

Especificaciones

(2Z)-3-[3-[3,5-Bis(trifluoromethyl)phenyl]-1H-1,2,4-triazol-1-yl]-2-propenoic acid 2-(2-pyrazinyl)hydrazide
1393477-72-9
Antiviral Compounds
C17H11F6N7O
Videos
Seguridad y manipulación

Seguridad y manipulación

Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

SDS
Documentos

Documentos

Certificados