missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma Semi-Micro Cuvettes, 104-QS (pair)
Highly pure Suprasil™ quartz glass cuvettes with PTFE lids.
Brand: Hellma 104-10-K-40
This item is not returnable.
View return policy
Description
- High-purity synthetic fused silica quartz construction
- UV/VIS range
- Absorption cell
- Includes PTFE lids
Specifications
1.4 mL | |
PTFE | |
PTFE lids | |
10 mm |
Quartz Suprasil | |
Layer thickness: 10 mm | |
2/Pk, Matched pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction