missing translation for 'onlineSavingsMsg'
Learn More

Hellma Semi-Micro Cuvettes, 108-QS (pair)

Highly pure Suprasil™ quartz glass cuvettes with PTFE lids.

Brand:  Hellma 108-000-10-K-40

Product Code. 12709166

  • 779.00 € / Pack of 2
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-purity synthetic fused silica quartz construction
  • UV/VIS range
  • PTFE lids included
Specifications

Specifications

1000 μL
PTFE
9 mm
2/Pk
200 to 2500 nm
12.5 mm
Suprasil Quartz
4 mm
PTFE Lids
10 mm
45 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.